Self-Standing Vials, with Caps
Supplier: NEST SCIENTIFIC
633002N 633012N 634002N 634012N 635002N 635012N
77508-488CS 472.55 USD
77508-488 77508-490 77508-492 77508-494 77508-500 77508-502
Self-Standing Vials, with Caps
Vials
This style of vial is has well standardized dimensions among most manufacturers, which makes them ideal for use with existing tube racks and automated filling equipment. They also feature knurling. Each vial also features an integrated silicone O-ring to further reduce risk of leaks.
  • Self- standing
  • O-ring seal
  • Sterile
  • External thread
  • Knurling for automation and easy grip
  • Suitable for a wide range of temperatures from −196 to +121 °C
  • Ideal for boiling applications
E-Beam sterilized and lot tested to SAL 10⁻⁶ to ISO 11137
Lot tested free DNase/RNase and DNA
Lot tested free BSE/TSE
Certified free of animal products
USP grade VI polypropylene
USP grade VI silicone O-ring
Nest scientific screw cap microcentrifuge tubes are a convenient and cost effective way to store small volumes of liquid samples securely without leaks. They are made of pure, virgin polypropylene with low levels of leachables, ideal for sensitive samples.
Certificaciones: Produced in ISO 13485:2016 and ISO 9001 facility. Molded and packed in ISO 7 cleanroom. Lot tested free of pyrogens USP 85.