2,4-Dimethylpyrrole ≥98% (by GC)

Supplier: Ambeed
Warning

Synonyms: 2,4-Dimethyl-1H-pyrrole

A993579-10G A993579-1G A993579-25G A993579-5G
77552-834EA 32 USD
77552-834 77552-836 77552-838 77552-840
2,4-Dimethylpyrrole ≥98% (by GC)
2,4-Dimethylpyrrole

Precaución: for Research Use Only

Formula: C₆H₉N
MDL Number: MFCD00192088
CAS Number: 625-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR