MAL-PEG8-NHS ester ≥95%

Supplier: Ambeed
Warning

A985907-1G A985907-100MG A985907-250MG
77552-820EA 495 USD
77552-820 77614-706 77687-007
MAL-PEG8-NHS ester ≥95%
MAL-PEG8-NHS ester

Precaución: for Research Use Only

Formula: C₃₀H₄₇ N₃O₁₅
MDL Number: MFCD11041144
CAS Number: 756525-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR