Tris(diethylamino)phosphine ≥98% (by GC)

Supplier: Ambeed
Danger

Synonyms: Hexaethyltriaminophosphine, Hexaethylphosphorus triamide, Hexaethyl phosphorous triamide

A984466-25G A984466-5G
77552-818EA 16 USD
77552-818 77599-350
Tris(diethylamino)phosphine ≥98% (by GC)
Tris(diethylamino)phosphine

Precaución: for Research Use Only

Formula: C₁₂H₃₀N₃P
MDL Number: MFCD00009041
CAS Number: 2283-11-6
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR