Sodium phosphate ≥96%

Supplier: Ambeed
Warning

Synonyms: tri-Sodium phosphate, Sodium phosphate tribasic, Trisodium phosphate

A983959-500G A983959-100G A983959-25G A983959-1000G
77552-816EA 8 USD
77552-816 77573-936 77573-938 77873-403
Sodium phosphate ≥96%
Sodium phosphate

Precaución: for Research Use Only

Formula: Na₃O₄P
MW: 163.94 g/mol
MDL Number: MFCD00003510
CAS Number: 7601-54-9
Merck Index: 12,08808

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR