(3S)-1,3-Dimethylpiperazin-2-one 95%

Supplier: Ambeed
Warning

A983494-5G A983494-250MG A983494-1G A983494-100MG
77552-814EA 1791 USD
77552-814 77569-704 77784-509 77784-508
(3S)-1,3-Dimethylpiperazin-2-one 95%
(3S)-1,3-Dimethylpiperazin-2-one

Precaución: For research use only.

Formula: C₆H₁₂N₂O
MDL Number: MFCD22201141
CAS Number: 1240300-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR