2,2'-Bis(2-oxazoline) ≥97%

Supplier: Ambeed
Warning

A980516-1G A980516-5G A980516-100G A980516-25G
77552-808EA 30.9 USD
77552-808 77573-928 77747-734 77747-735
2,2'-Bis(2-oxazoline) ≥97%
2,2'-Bis(2-oxazoline)

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
MDL Number: MFCD00059930
CAS Number: 36697-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR