4-(N,N-Dimethylamino)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Dimethylamino)benzeneboronic acid

A965786-1G A965786-5G A965786-10G A965786-250MG A965786-25G
77552-786EA 31 USD
77552-786 77569-692 77882-851 77882-852 77882-853
4-(N,N-Dimethylamino)phenylboronic acid 98%
4-(N,N-Dimethylamino)phenylboronic acid

Precaución: For research use only.

Formula: C₈H₁₂BNO₂
MDL Number: MFCD01074642
CAS Number: 28611-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR