1,3-Di-tert-butyl-5-iodobenzene ≥95%

Supplier: Ambeed
Warning

A958371-1G A958371-5G A958371-250MG
77552-776EA 35 USD
77552-776 77595-550 77749-790
1,3-Di-tert-butyl-5-iodobenzene ≥95%
1,3-Di-tert-butyl-5-iodobenzene

Precaución: for Research Use Only

Formula: C₁₄H₂₁I
MDL Number: MFCD16875483
CAS Number: 37055-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR