4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzylamine hydrochloride ≥98%

Supplier: Ambeed
Danger

Synonyms: 3-[4-(Aminomethyl)phenyl]-3-(trifluoromethyl)-3H-diazirine hydrochloride

A952055-1G A952055-250MG
77552-766EA 419 USD
77552-766 77552-768
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzylamine hydrochloride ≥98%
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzylamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₈F₃N₃·HCl
MDL Number: MFCD29765752
CAS Number: 1258874-29-1
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR