3-Fluoro-3-methylazetidine hydrochloride ≥98%

Supplier: Ambeed
Warning

A946555-5G A946555-1G A946555-250MG
77552-756EA 228 USD
77552-756 77573-882 77891-711
3-Fluoro-3-methylazetidine hydrochloride ≥98%
3-Fluoro-3-methylazetidine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₉ClFN
MDL Number: MFCD21605303
CAS Number: 1427379-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR