(Cyanomethyl)(trimethyl)phosphonium iodide ≥98%

Supplier: Ambeed
Warning

A942822-1G A942822-5G
77552-744EA 177 USD
77552-744 77552-746
(Cyanomethyl)(trimethyl)phosphonium iodide ≥98%
(Cyanomethyl)(trimethyl)phosphonium iodide

Precaución: for Research Use Only

Formula: C₅H₁₁NP·I
MDL Number: MFCD20483909
CAS Number: 42843-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR