Tris(2,2,2-trifluoroethyl)borate ≥98%

Supplier: Ambeed
Danger

A939525-25G A939525-1G
77552-742EA 157 USD
77552-742 77873-381
Tris(2,2,2-trifluoroethyl)borate ≥98%
Tris(2,2,2-trifluoroethyl)borate

Precaución: for Research Use Only

Formula: C₆H₆BF₉O₃
MDL Number: MFCD00093768
CAS Number: 659-18-7
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR