Potassium trichloroammineplatinate(II) ≥99.9% (trace metals basis)

Supplier: Ambeed
Warning

A932313-1G A932313-250MG A932313-100MG
77552-730EA 399 USD
77552-730 77569-654 77603-316
Potassium trichloroammineplatinate(II) ≥99.9% (trace metals basis)
Potassium trichloroammineplatinate(II)

Precaución: For research use only.

Formula: K(PtCl₃NH₃)
MDL Number: MFCD00145154
CAS Number: 13820-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR