tert-Butyl-3-(2-ethoxy-2-oxoethylidene)azetidine-1-carboxylate 96%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-(2-ethoxy-2-oxoethylidene)azetidine-1-carboxylate

A925110-25G A925110-5G
77552-718EA 323.42 USD
77552-718 77569-648
tert-Butyl-3-(2-ethoxy-2-oxoethylidene)azetidine-1-carboxylate 96%
tert-Butyl-3-(2-ethoxy-2-oxoethylidene)azetidine-1-carboxylate

Precaución: For research use only.

Formula: C₁₂H₁₉NO₄
MW: 241.29 g/mol
MDL Number: MFCD16140210
CAS Number: 1002355-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR