1-Amino-3,6,9,12-tetraoxapentadec-14-yne ≥97%

Supplier: Ambeed
Warning

Synonyms: Acetylene-PEG4-amine, 3,6,9,12-Tetraoxapentadec-14-yn-1-amine, Propargyl-PEG4-amine

A918870-1G A918870-250MG A918870-100MG
77552-714EA 62 USD
77552-714 77595-480 77623-144
1-Amino-3,6,9,12-tetraoxapentadec-14-yne ≥97%
1-Amino-3,6,9,12-tetraoxapentadec-14-yne

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₄
MW: 231.29 g/mol
MDL Number: MFCD22380736
CAS Number: 1013921-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR