(2R,3S)-3-Amino-2-hydroxy-4-phenylbutanoic acid ≥95%

Supplier: Ambeed
Warning

A914192-5G A914192-1G
77552-710EA 233.81 USD
77552-710 77552-708
(2R,3S)-3-Amino-2-hydroxy-4-phenylbutanoic acid ≥95%
(2R,3S)-3-Amino-2-hydroxy-4-phenylbutanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
MW: 195.21 g/mol
MDL Number: MFCD03411619
CAS Number: 62023-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR