1,3-Bis(3-aminopropyl)tetramethyldisiloxane ≥83%

Supplier: Ambeed
Danger

A902374-25G A902374-5G
77552-694EA 30 USD
77552-694 77747-666
1,3-Bis(3-aminopropyl)tetramethyldisiloxane ≥83%
1,3-Bis(3-aminopropyl)tetramethyldisiloxane

Precaución: for Research Use Only

Formula: C₁₀H₂₈N₂OSi₂
MDL Number: MFCD00039786
CAS Number: 2469-55-8
UN: 3267
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR