(1R,2R)-N1,N2-Dimethyl-1,2-bis(3-(trifluoromethyl)phenyl)ethane-1,2-diamine ≥97%

Supplier: Ambeed
Danger

A883908-1G A883908-250MG
77552-664EA 250.29 USD
77552-664 77573-784
(1R,2R)-N1,N2-Dimethyl-1,2-bis(3-(trifluoromethyl)phenyl)ethane-1,2-diamine ≥97%
(1R,2R)-N1,N2-Dimethyl-1,2-bis(3-(trifluoromethyl)phenyl)ethane-1,2-diamine

Precaución: for Research Use Only

Formula: C₁₈H₁₈F₆N₂
MW: 376.33 g/mol
MDL Number: MFCD00075560
CAS Number: 137944-39-9
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR