3-Fluorobicyclo[1.1.1]pentane-1-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A883164-1G A883164-250MG A883164-100MG A883164-5G
77552-662EA 164.8 USD
77552-662 77595-430 77599-208 77599-210
3-Fluorobicyclo[1.1.1]pentane-1-carboxylic acid ≥98%
3-Fluorobicyclo[1.1.1]pentane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₇FO₂
MW: 130.117 g/mol
MDL Number: MFCD19228680
CAS Number: 146038-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR