tert-Butyl 4-hydrazinylpiperidine-1-carboxylate oxalate ≥98%

Supplier: Ambeed
Warning

A882281-1G A882281-5G
77552-658EA 46.35 USD
77552-658 77595-428
tert-Butyl 4-hydrazinylpiperidine-1-carboxylate oxalate ≥98%
tert-Butyl 4-hydrazinylpiperidine-1-carboxylate oxalate

Precaución: for Research Use Only

Formula: C₁₂H₂₃N₃O₆
MW: 305.3275 g/mol
MDL Number: MFCD31695834
CAS Number: 2108550-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR