3,4-Dimethoxythiophenol ≥98%

Supplier: Ambeed
Danger

A878566-25G A878566-1G A878566-5G
77552-650EA 248 USD
77552-650 77573-774 77595-426
3,4-Dimethoxythiophenol ≥98%
3,4-Dimethoxythiophenol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂S
MDL Number: MFCD00052823
CAS Number: 700-96-9
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR