4-Bromo-1-butyne ≥97%

Supplier: Ambeed
Danger

A877480-1G A877480-25G A877480-5G
77552-644EA 15.45 USD
77552-644 77552-646 77552-648
4-Bromo-1-butyne ≥97%
4-Bromo-1-butyne

Precaución: for Research Use Only

Formula: C₄H₅Br
MW: 132.99 g/mol
MDL Number: MFCD10000883
CAS Number: 38771-21-0
UN: 1992
ADR: 3,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR