3-Fluoro-2-hydroxybenzeneboronic acid ≥96%

Supplier: Ambeed
Warning

Synonyms: 2-Borono-6-fluorophenol

A865394-5G A865394-1G A865394-250MG
77552-630EA 267.8 USD
77552-630 77573-746 77749-674
3-Fluoro-2-hydroxybenzeneboronic acid ≥96%
3-Fluoro-2-hydroxybenzeneboronic acid

Precaución: for Research Use Only

Formula: C₆H₆BFO₃
MW: 155.919 g/mol
MDL Number: MFCD07781238
CAS Number: 259209-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR