Octadecanal ≥98%

Supplier: Ambeed
Warning

Synonyms: Octadecyl Aldehyde, Octadecanaldehyde, Stearyl Aldehyde, Stearaldehyde

A860342-5G A860342-1G A860342-25G
77552-618EA 73.13 USD
77552-618 77684-821 77684-822
Octadecanal ≥98%
Octadecanal

Precaución: For research use only.

Formula: C₁₈H₃₆O
MW: 268.49 g/mol
MDL Number: MFCD03790007
CAS Number: 638-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR