N-Succinimidyl Bromoacetate ≥98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dioxopyrrolidin-1-yl 2-bromoacetate, N-(Bromoacetoxy)succinimide, Bromoacetic Acid N-Succinimidyl Ester, Bromoacetic acid N-hydroxysuccinimide ester

A850820-1G A850820-5G A850820-250MG A850820-25G A850820-100MG
77552-600EA 54 USD
77552-600 77573-700 77627-660 77627-662 77627-658
N-Succinimidyl Bromoacetate ≥98%
N-Succinimidyl Bromoacetate

Precaución: for Research Use Only

Formula: C₆H₆BrNO₄
MDL Number: MFCD00058571
CAS Number: 42014-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR