3-(3-(But-3-yn-1-yl)-3H-diazirin-3-yl)propanoic acid ≥95%

Supplier: Ambeed
Danger

Synonyms: 3-(3-(But-3-yn-1-yl)-3H-diazirin-3-yl)propan-1-amine

A849359-50MG A849359-25MG
77552-594EA 404.79 USD
77552-594 77573-696
3-(3-(But-3-yn-1-yl)-3H-diazirin-3-yl)propanoic acid ≥95%
3-(3-(But-3-yn-1-yl)-3H-diazirin-3-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
MDL Number: MFCD31433997
CAS Number: 1450754-37-6
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR