3-(Dimethylamino)acrolein ≥98%

Supplier: Ambeed
Danger

A848990-25G A848990-5G
77552-592EA 53 USD
77552-592 77749-656
3-(Dimethylamino)acrolein ≥98%
3-(Dimethylamino)acrolein

Precaución: for Research Use Only

Formula: C₅H₉NO
MDL Number: MFCD00006999
CAS Number: 927-63-9
UN: 3267
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR