Ethyl difluoroacetate ≥98%

Supplier: Ambeed
Danger

A842841-25G A842841-100G A842841-10G
77552-576EA 14 USD
77552-576 77670-208 77880-642
Ethyl difluoroacetate ≥98%
Ethyl difluoroacetate

Precaución: for Research Use Only

Formula: C₄H₆F₂O₂
MDL Number: MFCD00013578
CAS Number: 454-31-9
UN: 2924
ADR: 3,8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR