2,5-Dibromo-4-methyl-3-nitropyridine ≥98%

Supplier: Ambeed
Warning

A827873-1G A827873-5G
77552-544EA 85 USD
77552-544 77595-316
2,5-Dibromo-4-methyl-3-nitropyridine ≥98%
2,5-Dibromo-4-methyl-3-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₄Br₂N₂O₂
MW: 295.93 g/mol
MDL Number: MFCD09839233
CAS Number: 947534-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR