1-Fluoro-3,3-dimethyl-1,2-benziodoxole ≥98%

Supplier: Ambeed
Danger

A826719-1G A826719-5G A826719-250MG A826719-100MG
77552-530EA 85.49 USD
77552-530 77552-532 77599-142 77749-618
1-Fluoro-3,3-dimethyl-1,2-benziodoxole ≥98%
1-Fluoro-3,3-dimethyl-1,2-benziodoxole

Precaución: for Research Use Only

Formula: C₉H₁₀FIO
MW: 280.08 g/mol
MDL Number: MFCD28397076
CAS Number: 1391728-13-4
UN: 1479
ADR: 5.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR