2,6-Bis(trifluoromethyl)benzonitrile ≥98%

Supplier: Ambeed
Warning

A825179-1G A825179-5G
77552-524EA 44 USD
77552-524 77900-716
2,6-Bis(trifluoromethyl)benzonitrile ≥98%
2,6-Bis(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₉H₃F₆N
MDL Number: MFCD00061226
CAS Number: 25753-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR