1-Methylcyclohexene 97%

Supplier: Ambeed
Danger

A818838-1G A818838-5G
77552-512EA 11 USD
77552-512 77569-522
1-Methylcyclohexene 97%
1-Methylcyclohexene

Precaución: For research use only.

Formula: C₇H₁₂
MDL Number: MFCD00001548
CAS Number: 591-49-1
UN: 3295
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR