2-Oxaspiro[3.3]heptan-6-amine hydrochloride ≥97%

Supplier: Ambeed
Warning

A813225-1G A813225-100MG A813225-250MG
77552-504EA 333 USD
77552-504 77886-800 77886-801
2-Oxaspiro[3.3]heptan-6-amine hydrochloride ≥97%
2-Oxaspiro[3.3]heptan-6-amine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁NO·ClH
MDL Number: MFCD22415297
CAS Number: 1523618-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR