(S)-2-Amino-2-(3,5-di-tert-butylphenyl)ethanol ≥95%

Supplier: Ambeed
Warning

A808355-1G A808355-100MG
77552-492EA 362.56 USD
77552-492 77622-968
(S)-2-Amino-2-(3,5-di-tert-butylphenyl)ethanol ≥95%
(S)-2-Amino-2-(3,5-di-tert-butylphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₆H₂₇NO
MW: 249.3917 g/mol
MDL Number: MFCD09253894
CAS Number: 1213667-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR