Methyl-2-(bromomethyl)acrylate ≥98% stabilised

Supplier: Ambeed
Danger

Synonyms: 2-(Bromomethyl)acrylic Acid Methyl Ester, Methyl 2-(bromomethyl)acrylate

Stabilization: Stabilized with BHT or HQ

A803846-25G A803846-5G A803846-1G
77552-472EA 315 USD
77552-472 77880-599 77880-598
Methyl-2-(bromomethyl)acrylate ≥98% stabilised
Methyl-2-(bromomethyl)acrylate

Precaución: for Research Use Only

Formula: C₅H₇BrO₂
MW: 179.01 g/mol
MDL Number: MFCD00011697
CAS Number: 4224-69-5
UN: 3265
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR