(4-Cyclopropyl-6-methoxypyrimidin-5-yl)boronic acid ≥97%

Supplier: Ambeed
Warning

A801940-25G A801940-5G A801940-1G
77552-466EA 276.04 USD
77552-466 77749-586 77749-585
(4-Cyclopropyl-6-methoxypyrimidin-5-yl)boronic acid ≥97%
(4-Cyclopropyl-6-methoxypyrimidin-5-yl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BN₂O₃
MW: 193.995 g/mol
MDL Number: MFCD31556945
CAS Number: 1798304-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR