1,4-Oxazepane ≥98%

Supplier: Ambeed
Danger

Synonyms: Homomorpholine

A799589-25G A799589-5G A799589-1G A799589-250MG
77552-456EA 410 USD
77552-456 77573-554 77884-739 77884-740
1,4-Oxazepane ≥98%
1,4-Oxazepane

Precaución: for Research Use Only

Formula: C₅H₁₁NO
MW: 101.1 g/mol
MDL Number: MFCD09971804
CAS Number: 5638-60-8
UN: 2920
ADR: 8,3 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR