2,2'-Oxybis(N,N-di-n-octylacetamide) 95%

Supplier: Ambeed
Warning

A798562-5G A798562-25G A798562-1G
77552-450EA 96 USD
77552-450 77569-494 77886-779
2,2'-Oxybis(N,N-di-n-octylacetamide) 95%
2,2'-Oxybis(N,N-di-n-octylacetamide)

Precaución: For research use only.

Formula: C₃₆H₇₂N₂O₃
MDL Number: MFCD23135286
CAS Number: 342794-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR