tert-Butyl (2-iodoethyl)carbamate ≥95%

Supplier: Ambeed
Danger

A793539-1G A793539-250MG A793539-5G
77552-446EA 77 USD
77552-446 77614-420 77886-766
tert-Butyl (2-iodoethyl)carbamate ≥95%
tert-Butyl (2-iodoethyl)carbamate

Precaución: for Research Use Only

Formula: C₇H₁₄INO₂
MW: 271.09 g/mol
MDL Number: MFCD09951818
CAS Number: 122234-46-2
UN: 1759
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR