3-Iodo-1-methylpyridin-2(1H)-one ≥97%

Supplier: Ambeed
Warning

A790919-250MG A790919-1G
77552-440EA 400.67 USD
77552-440 77573-530
3-Iodo-1-methylpyridin-2(1H)-one ≥97%
3-Iodo-1-methylpyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₆H₆INO
MW: 235.224 g/mol
MDL Number: MFCD23768850
CAS Number: 615534-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR