2,2,2-Trifluoroethyl formate ≥98%

Supplier: Ambeed
Danger

A786716-25G A786716-5G
77552-434EA 108 USD
77552-434 77618-178
2,2,2-Trifluoroethyl formate ≥98%
2,2,2-Trifluoroethyl formate

Precaución: for Research Use Only

Formula: C₃H₃F₃O₂
MW: 128.05 g/mol
MDL Number: MFCD06797733
CAS Number: 32042-38-9
UN: 1993
ADR: 3 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR