7-Bromo-2,4,6-trichloro-8-fluoroquinazoline ≥98%

Supplier: Ambeed
Warning

A778703-5G A778703-1G
77552-412EA 327.54 USD
77552-412 77573-506
7-Bromo-2,4,6-trichloro-8-fluoroquinazoline ≥98%
7-Bromo-2,4,6-trichloro-8-fluoroquinazoline

Precaución: for Research Use Only

Formula: C₈HBrCl₃FN₂
MW: 330.368 g/mol
MDL Number: MFCD29918562
CAS Number: 1698028-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR