Methyl trifluoromethanesulfonate ≥97%

Supplier: Ambeed
Danger

A777184-10G A777184-25G A777184-5G
77552-400EA 23.69 USD
77552-400 77552-402 77552-404
Methyl trifluoromethanesulfonate ≥97%
Methyl trifluoromethanesulphonate

Precaución: for Research Use Only

Formula: C₂H₃F₃O₃S
MW: 164.11 g/mol
MDL Number: MFCD00000409
CAS Number: 333-27-7
UN: 2920
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR