2-Ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Danger

Synonyms: Ethynylboronic acid Pinacol ester

A777138-1G A777138-5G
77552-398EA 15 USD
77552-398 77569-472
2-Ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-Ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: For research use only.

Formula: C₈H₁₃BO₂
MDL Number: MFCD13182122
CAS Number: 347389-74-6
UN: 1325
ADR: 4.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR