tert-Butyl 4-(piperidin-3-ylmethyl)piperazine-1-carboxylate dihydrochloride ≥98%
Supplier: Ambeed
|
Warning
|
A775718-1G
A775718-250MG
A775718-100MG
77552-394EA
227
USD
77552-394
77599-066
77622-926
tert-Butyl 4-(piperidin-3-ylmethyl)piperazine-1-carboxylate dihydrochloride ≥98%
tert-Butyl 4-(piperidin-3-ylmethyl)piperazine-1-carboxylate dihydrochloride
Precaución: for Research Use Only
Formula:
C₁₅H₃₁Cl₂N₃O₂ |
MDL Number:
MFCD20526295 CAS Number: 1211498-87-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...