1,2-Epoxycyclopentane ≥98%

Supplier: Ambeed
Danger

Synonyms: Cyclopentene oxide

A767801-25G A767801-5G A767801-100G
77552-364EA 107.12 USD
77552-364 77552-366 77747-520
1,2-Epoxycyclopentane ≥98%
1,2-Epoxycyclopentane

Precaución: for Research Use Only

Formula: C₅H₈O
MW: 84.12
MDL Number: MFCD00005161
CAS Number: 285-67-6
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR