1-Methyl-1-propylpyrrolidinium bis(fluorosulfonyl)imide ≥98%

Supplier: Ambeed
Warning

A765358-25G A765358-5G
77552-356EA 122 USD
77552-356 77622-916
1-Methyl-1-propylpyrrolidinium bis(fluorosulfonyl)imide ≥98%
1-Methyl-1-propylpyrrolidinium bis(fluorosulfonyl)imide

Precaución: for Research Use Only

Formula: C₈H₁₈F₂N₂O₄S₂
MW: 308.36 g/mol
MDL Number: MFCD17018948
CAS Number: 852620-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR