3-(2-Pyridyldithio)propionic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: 3-(Pyridin-2-yldisulphanyl)propanoic acid, 3-(2-Pyridyldithio)propanoic acid, PDTP

A763639-1G A763639-5G A763639-250MG A763639-100MG A763639-25G
77552-342EA 134.93 USD
77552-342 77552-344 77618-104 77686-773 77686-774
3-(2-Pyridyldithio)propionic acid ≥97%
3-(2-Pyridyldithio)propionic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₂S₂
MW: 215.29 g/mol
MDL Number: MFCD18252554
CAS Number: 68617-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR