N-Boc-peg4-bromide 98%

Supplier: Ambeed
Warning

A763086-1G A763086-5G A763086-250MG A763086-100MG
77552-340EA 67 USD
77552-340 77569-452 77599-042 77622-896
N-Boc-peg4-bromide 98%
N-Boc-peg4-bromide

Precaución: For research use only.

Formula: C₁₅H₃₀BrNO₆
MDL Number: MFCD31536756
CAS Number: 1392499-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR